Learn More
Abeba™ Crawler ALU 34501 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Marca:  Abeba™ 34501/47
Dettagli aggiuntivi : Peso : 1.24000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15619867
  • 101.00€ / 1 paio
Spedizione stimata
25-11-2020
Aggiungi al carrello
Descrizione
Descrizione
  • CE EN ISO 20345:2011 S2 SRC
  • Microfiber black/blue
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Upper one piece
Specifica
Specifica
CE EN ISO 20345:2011 S2 SRC
Microfiber
Slip-resistant
34501
Black
47
Polyurethane
Aluminum