Learn More
Abeba™ Crawler ALU 34570 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Marca:  Abeba™ 34570/36
Additional Details : Peso : 1.23200kg
 View more versions of this product
Product Code. 15619947
  • 106.00€ / 1 paio
Spedizione stimata
30-11-2020
Add to basket
Description
Description
  • CE EN ISO 20345:2011 S1 SRC
  • Smooth leather with breathable textile inlays
  • White
  • Aluminum toe cap
  • Foamed PU midsole
  • Slip resistance TPU outsole sole
  • Replaceable acc Wave insole (article no. 3577)
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Reflective material
  • Discontinued model
  • Scratch-resistant coating in toe and heel area
Specifications
Specifications
CE EN ISO 20345:2011 S1 SRC
Leather
Slip-resistant
34570
White
36
Polyurethane
Aluminum
Videos
SDS
Documents