Learn More
Macherey-Nagel™ Crimp Closures N8 with Hole
Product Code. 15855421 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Silver
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15855421 Silver Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15855421 Supplier Macherey-Nagel™ Supplier No. 702025
In stock
Aluminum crimp N8 closures have a red rubber/PTFE septa material for routine analysis. All closures are automatically assembled under strict hygienic conditions. They are compitable with Dionex, Perkin Elmer and Thermo Scientific autosamplers.
  • Red rubber/PTFE septa
  • N8 Diameter
  • Ideal for GC and HPLC applications
Specifications
Color (Seal) Red-Orange/Colorless
Description Aluminum Crip Cap with Hole, Prefitted Septa
Color Silver
Material Aluminum
Septum Red Rubber/PTFE
For Use With (Application) GC, HPLC
Temperature Range (Metric) Temp. Resistance -40°C up to 110°C/Analytical Purity - Medium/Medium Hard to Penetrate/Medium Resealability
Thread Style N8
Thickness (Metric) 1.0 mm (Septa)
For Use With (Equipment) Dionex, PerkinElmer, Thermo Scientific Autosamplers
Product Type Crimp Seal Vial