missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cas9 Monoclonal Antibody (7A9-3A3)
GREENER_CHOICE

Codice prodotto. 15658018
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15658018 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15658018 Fornitore Invitrogen™ N. del fornitore MA1201

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

Cas9 (7A9-3A3) Mouse mAb recognizes transfected levels of total Cas9 protein. Product was tested in Streptococcus pyogenes bacteria.

Cas9 is an RNA-guided DNA endonuclease enzyme associated with the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) adaptive immunity system in bacteria and genetic engineering. The system is unique and flexible due to its dependence on RNA as the element that targets the nuclease to a desired DNA sequence. It can be used to induce indel mutations, specific sequence replacements/insertions, and large deletions or genomic rearrangements at any desired location in the genome. In addition, Cas9 can also be used to mediate upregulation of specific endogenous genes or to alter histone modifications or DNA methylation. Cas9 may be derived from Streptococcus pyogenes (SpCas9) or Staphylococcus aureus (SaCas9). SaCas9 has a number of properties that make it advantageous for genome editing, including its small size, high efficiency, nickase activity, and apparent specificity. SaCas9 principally recognizes an NNGRRT protospacer adjacent motif (PAM) and cleaves target DNA at rates comparable to SpCas9 with an ideal spacer lengths of 24 to 20 nucleotides. In comparison to SpCas9, SaCas9 is smaller (1368 AA versus 1053 AA) and generates indels at a comparable rate when directed against target DNA with the mutually permissive NGGRRT PAM. This makes it the preferred enzyme for all-in-one delivery of Cas9 and multiple gRNA expression cassettes with Adeno-associated vectors.
TRUSTED_SUSTAINABILITY

Specifica

Antigene Cas9
Applicazioni Western Blot, Immunocytochemistry
Classificazione Monoclonal
Clone 7A9-3A3
Concentrazione 1 mg/mL
Coniugato Unconjugated
Formulazione PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Gene cas9
N. accesso geni Q99ZW2
Alias gene cas9; CRISPR9; CRISPR-Cas9; s aureus
Simboli geni cas9
Specie ospite Mouse
Immunogeno Cas9 nuclease.
Metodo di purificazione Protein G
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 901176
Target Species Bacteria
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Forma Liquid
Isotype IgG1 κ
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.