Learn More
Cytiva Cross Linked Dextran Matrix Cytodex 1
Codice prodotto. 11319981 Sfoglia Tutto Cytiva Prodotti
Cambia vista
Click to view available options
Quantità:
100 g
25 g
500 g
Dimensione della confezione:
25g
500g
Pezzo
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11319981 25 g 25g
11389971 100 g Pezzo
14801611 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 11319981 Fornitore Cytiva N. del fornitore 17044801
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello
Aggiungi al carrello
Cytodex 1 is a general purpose microcarrier. It is particularly suitable for most established cell lines and for production of viruses or cell products from cultures of primary cells and normal diploid cell strains. Microcarrier culture is a technique which makes possible the practical high yield culture of anchorage-dependent cells. Cytodex has been specifically developed for the culture of a wide range of animal cells in culture volumes ranging from a few milliliters to more than six thousand liters. Using Cytodex in simple suspension culture systems provides yields of several million cells per milliliter. Formed by substituting a cross-linked dextran matrix with positively charged DEAE groups distributed throughout the matrix, is a general purpose microcarrier.
  • General-purpose microcarrier for most established, anchorage-dependent cell lines.
  • Contains positive charges throughout the microporous matrix.
  • Transparent for easy microscopic examination of attached cells.
TRUSTED_SUSTAINABILITY
Specifica
Product Type Cytodex 1
Quantità 25 g
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.