Learn More
CROT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA553906
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: LAKSTEERTF QYQDSLPSLP VPSLEESLKK YLESVKPFAN QEEYKKTEEI VQKFQSGIGE KLHQKLLERA KGKRNWLEEW WLNVAYLDVR IPSQLNVNFA GPAAHFEHYW PPKEGTQLER GSITLWH Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
CROT Polyclonal antibody specifically detects CROT in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifica
CROT | |
Polyclonal | |
Unconjugated | |
Crot | |
1200003H03Rik; carnitine octanoyltransferase; carnitine O-octanoyltransferase; COT; CROT; CROT protein; hypothetical protein LOC553204; peroxisomal carnitine acyltransferase; peroxisomal carnitine O-octanoyltransferase; zgc:110643 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54677 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UKG9 | |
Crot | |
Recombinant protein corresponding to Human CROT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only