Learn More
BD Biosciences™ CS&T Beads (CE/IVD)
Codice prodotto. 15816349
Cambia vista
Click to view available options
N. di test:
150 Tests
50 Tests
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. N. di test unitSize
15816349 150 Tests Pezzo
15806349 50 Tests Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15816349 Fornitore BD Biosciences™ N. del fornitore 656505
 Articolo soggetto a restrizioni
Spedizione stimata: 21-08-2026
Aggiungi al carrello
Aggiungi al carrello
Provide a standardized method to perform quality control of the instrument’s optics, electronics, and fluidics, and for adjusting fluorescence compensation.
  • Used on a BD flow cytometer.
  • Provides a standardized method to perform quality control of the instrument's optics, electronics, and fluidics, and for adjusting fluorescence compensation.
  • On some BD instruments, BD CS&T beads are also used for adjusting detector voltages.
  • Suspension of fluorospheres with uniform and stable size and fluorescence intensity.
  • Used for instrument quality control (QC) to characterize, track, and report performance measurements of supported flow cytometers.
  • Dyed with fluorochromes that are excited by the cytometer's lasers.
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage Three 3 mL vials at 50 tests per vial for 150 tests per kit
N. di test 150 Tests
Stabilità The beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. After dilution, the beads are stable for 8 hours at 2°C–25°CC
Product Type Fluorescent Polystyrene Beads