Learn More
BD Biosciences™ CS&T IVD Beads (CE/IVD)
Product Code. 15856339 Shop All BD Biosciences Products
Change view
Click to view available options
No. of Tests:
150 Tests
50 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests Unit Size
15856339 150 Tests Each
15846339 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15856339 Supplier BD Biosciences™ Supplier No. 656047
 Restricted Item
Spedizione stimata: 03-12-2026
Used to set up your cytometer, perform daily quality control, and determine lyse/wash application settings.
  • Intended for in vitro diagnostic use on BD FACSCanto™ II flow cytometers running BD FACSDiva™ software.
  • Used to set up the cytometer, to perform daily performance quality control (QC), and to determine lyse/wash (LW) application settings.
  • Dyed with fluorochromes that are excited by the cytometer's lasers.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Three 3 mL vials at 50 tests per vial for 150 tests per kit
No. of Tests 150 Tests
Stability The beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. After dilution, the beads are stable for 8 hours at 2°C–25°CC
Product Type Fluorescent Polystyrene Beads