Descrizione
- Designed for use on BD™ flow cytometers running FACSuite™ software.
- Enable the software to automatically run performance QC (daily) and characterization GC (every 6 months).
- Dyed with fluorochromes that are excited by the cytometer's lasers.
- Beads emit fluorescence in detectors.
Specifica
Specifica
| Content And Storage | Store vials at 2°C to 8°C and protect from light. Do not freeze. |
| Excitation/Emission | Blue laser: 500-800 nm, Red laser: 650-800 nm, Violet laser: 420-700 nm |
| N. di test | 25 tests/vial, 50 tests/kit |
| Quantità | 2 x 3 mL vials |
| Stabilità | Beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. |