Learn More
behr Labor-Technik™ CSB 12/E Precision Heating Block
Descrizione
- Casing made of powder-coated steel and non-corrosive stainless steel
- Extensive insulation of the heating block for unobjectionable temperatures on the casing surface
- End-to-end grooves in the plates at the bottom of the bore facilitate sample handling and increase the safety of the user
- During insertion of the reaction vessels, the grooves balance the pressure; this way, the vessels can be lowered precisely into the bores and the user can work with the insert frames without any problems
- Traces of liquid on the outer walls of the reaction vessels evaporate immediately when inserting the sample in the hot block; the grooves guide the steam off and prevent the breakage of glass as a result of them jumping from the pressure
- In case the vessel overflows, the sample flows outward via the groove, reducing the risk of acid damages in the heating block
- Extensive construction guarantees an exact and even temperature at all sample positions
Specifica
Specifica
| Descrizione | Precision heating block thermostat for 12 reaction vessels RG, 20°C to 300°C |
| No. of Places | 12 Sample Positions |
| N. di test | 12 Sample Positions |
| Quantità | 1 Ea. |
| Tipo | Precision Heating Block |