Learn More
CleanSpace™ Technology CST Cleaning & Storage Plug
Product Code. 17855085 Shop All CleanSpace Technology Products
Change view
Click to view available options
Product Type:
Cleaning and Storage Plug
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
17855085 Cleaning and Storage Plug Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17855085 Supplier CleanSpace™ Technology Supplier No. CST1024
  • ONLINE EXCLUSIVE PRICE
    74.76€
    58.00€ / pezzi 1
    Risparmia 16.76€
    22% Sconto
Spedizione stimata: 07-01-2027
The CleanSpace CST Cleaning and Storage Plug is designed to be used only with the CleanSpace CST ULTRA Power Unit (CST1012).
  • Designed to be used only with the CleanSpace CST ULTRA Power Unit (CST1010)
  • With the filter removed from the CleanSpace CST ULTRA Power Unit, this accessory is used to seal all openings (both bellows and filter air inlet).
  • During cleaning, all openings must be sealed.
  • It is also recommended that all openings are sealed and after cleaning and decontamination, and during storage.
Specifications
Product Type Cleaning and Storage Plug