Learn More
Thermo Scientific™ Culti-Loops™ Bacteroides ovatus ATCC™ BAA-1296™
Codice prodotto. 13226179
Cambia vista
Click to view available options
Quantità:
5 Loops/Pack
Dimensione della confezione:
pezzi 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
13226179 5 Loops/Pack pezzi 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 13226179 Fornitore Thermo Scientific™ N. del fornitore R4609374
Please to purchase this item. Need a web account? Register with us today!
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifica
ATCC® BAA-1296™*
Descrizione Bacteroides ovatus
Formato Culti-Loops
Product Type Bacteroides species
Target Organism Class Anaerobes
Test Type Qualitative QC
Quantità 5 Loops/Pack
Strain Designations Vitek 400963; NSB 24257
Specie Bacteroides species
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.