Learn More
Thermo Scientific™ Culti-Loops™ Sporothrix schenckii ATCC™ 10212™
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifica
Specifica
| ATCC® | 10212TM* |
| Descrizione | Sporothrix schenckii ATCC™ 10212™ |
| Product Type | Culti-Loops Quality Control Organism |
| Target Organism Class | Fungi |
| Test Type | Qualitative QC |
| Tipo di imballaggio | 5 Loops/Package |
| Strain Designations | NIH 7019 |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.