Learn More
R&D Systems™ Cultrex Antibiotic-free Mouse Laminin I, Pathclear
Codice prodotto. 18775588
Cambia vista
Click to view available options
Quantità:
1 mg
Dimensione della confezione:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18775588 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18775588 Fornitore R&D Systems™ N. del fornitore 340101002
  • ONLINE EXCLUSIVE PRICE
    272.00€
    257.25€ / 1mg
    Risparmia 14.75€
    5% Sconto
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello
Aggiungi al carrello
Supports the attachment of MG63 osteosarcoma cells at <20 μg/mL.
Sterility Testing:
  • PathClear™ - Negative by PCR test for mycoplasma; 17 bacterial and virus strains typically included in mouse antibody production (MAP) testing, plus 13 additional murine infectious agents including LDEV, for a total of 31 organisms and viruses.
  • No bacterial or fungal growth detected after incubation at 37°C for 14 days following USP sterility testing guidelines.
  • Endotoxin concentration < 20 EU/mL by LAL assay.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 1 mg
Sinonimo LAMA, LAMA1, laminin subunit alpha 1, Laminin1, Laminin-111, PTBHS, S-LAM-alpha