Learn More
Custom Primer
Codice prodotto. 10358349
Codice prodotto. 10358349 Fornitore Invitrogen N. del fornitore 11732054
 Articolo soggetto a restrizioni
Spedizione stimata: 15-10-2026
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole.
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).