Learn More
WLD Tec™ Safety Adapter for CV360 Gas Cartridge
Product Code. 10171473
Change view
Click to view available options
For Use With (Equipment):
CV360 Gas Cartridge
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10171473 CV360 Gas Cartridge Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10171473 Supplier WLD Tec™ Supplier No. 8.000.700
  • ONLINE EXCLUSIVE PRICE
    152.56€
    115.00€ / pezzi 1
    Risparmia 37.56€
    24% Sconto
Spedizione stimata: 11-12-2026
Add to basket
Add to basket
Suitable and safe gas cartridge adapter for Gasprofi 1 SCS laboratory gas burners. WLD Tec™ Safety Adapter for CV360 Gas Cartridge includes and integrated pressure regulator.
  • Safety adapter
  • Includes integrated pressure regulator
  • Accepts gas cartridges, CV 360
Specifications
For Use With (Equipment) CV360 Gas Cartridge
Includes 50 mbar Integrated Pressure Regulator
Product Type Gas Adapter