missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Molecular weight: 247.29 Da
- Purity: >99%
- Description: AMPA receptor potentiator that selectively binds to the agonist bound non-desensitized receptor.
- Function: Destabilizes desensitized receptor conformation with greater potency than CX516 and enhances cognitive function in vivo.
- Form: Solid
- Storage: Store at -20°C.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| Forma | Solid |
| Shipping Condition | Ambient |
| Target Molecule | AMPA Receptor |
| Formulazione | 100% CX 546 |
| Quantità | 50 mg |
| Da utilizzare con (applicazione) | AMPA receptor potentiation, Cognitive function research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?