missing translation for 'onlineSavingsMsg'
Learn More

Cyclin B1 Antibody (CCNB1/9242R), mFluor Violet 450 SE, Novus Biologicals™

Codice prodotto. 18555937 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18555937 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18555937 Fornitore Novus Biologicals N. del fornitore NBP324309MFV450

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Monoclonal Antibody

Cyclin B1 Monoclonal antibody specifically detects Cyclin B1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Spezifikation

Antigene Cyclin B1
Applicazioni Immunohistochemistry (Paraffin)
Classificazione Monoclonal
Clone CCNB1/9242R
Coniugato mFluor Violet 450 SE
Formulazione 50 mM Sodium Borate
Alias gene CCNB, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1
Specie ospite Rabbit
Immunogeno His-tagged recombinant hamster Cyclin B1 protein
Metodo di purificazione Protein A or G purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Cell Cycle and Replication, Cytokine Research, Immunology, Mesenchymal Stem Cell Markers, Mitotic Regulators, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Stem Cell Lines, Stem Cell Markers, Tumor Suppressors
Primario o secondario Primary
ID gene (immissione) 891
Target Species Human
Content And Storage Store at 4°C in the dark.
Forma Purified
Isotype IgG κ
Mehr anzeigen Weniger anzeigen
Name des Produkts
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.