Learn More
Hellma™ Cylindrical cell far UV quartz 0.1mm light path 160μL Hellma
Product Code. 12721705
Change view
Click to view available options
Lightpath:
0.1 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lightpath Unit Size
12721705 0.1 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12721705 Supplier Hellma™ Supplier No. 1210.1040
  • ONLINE EXCLUSIVE PRICE
    535.00€
    487.00€ / pezzi 1
    Risparmia 48.00€
    8% Sconto
Spedizione stimata: 02-12-2026