Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.
Marca:  Lenz Laborglasinstrumente™ B0496271
Dettagli aggiuntivi : Peso : 0.10000kg
Codice prodotto. 10020720
  • 205.00€ / Pezzo
Spedizione stimata: 31-05-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • With inscription label
  • With NS socket and cone
  • With scale (approx. volume/mL)
  • With pressure equalizing tube
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specifica
Specifica
1000 mL
Duran Glass, Borosilicate Glass