Learn More
Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.
Marca: Lenz Laborglasinstrumente™ B0496441
Dettagli aggiuntivi : Peso : 0.10800kg
Descrizione
- With inscription label
- With NS socket and cone
- With scale (approx. volume/mL)
- With pressure equalizing tube
- With screwthread retaining device
- Corrosion-free and resistant to thermal shock
Specifica
25 mL | |
Duran Glass, Borosilicate Glass | |
Pressure Equalization |