Learn More
Lenz Laborglasinstrumente™ Imbuti contagocce cilindrici con tappo in vetro
Codice prodotto. 10539241
Cambia vista
Click to view available options
Capacità (sistema metrico):
25 mL
50 mL
100 ml
250 mL
500 mL
1000 mL
Cone Size:
14/23 NS
29/32 NS
Include:
Pressure Equalizing Tube
w/o Pressure Equalizing Tube
Dimensione della confezione:
pezzi 2
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) Cone Size Include unitSize
10539241 50 mL 29/32 NS w/o Pressure Equalizing Tube pezzi 2
10294431 1000 mL 29/32 NS w/o Pressure Equalizing Tube pezzi 2
10403881 1000 mL 29/32 NS Pressure Equalizing Tube pezzi 2
10616331 100 ml 29/32 NS Pressure Equalizing Tube pezzi 2
10632551 100 ml 29/32 NS w/o Pressure Equalizing Tube pezzi 2
10303291 100 ml 14/23 NS w/o Pressure Equalizing Tube pezzi 2
10484631 250 mL 29/32 NS w/o Pressure Equalizing Tube pezzi 2
10313101 250 mL 29/32 NS Pressure Equalizing Tube pezzi 2
10210841 25 mL 14/23 NS w/o Pressure Equalizing Tube pezzi 2
10139320 50 mL 14/23 NS Pressure Equalizing Tube pezzi 2
10673891 500 mL 29/32 NS Pressure Equalizing Tube pezzi 2
10644661 50 mL 14/23 NS w/o Pressure Equalizing Tube pezzi 2
10557321 50 mL 29/32 NS Pressure Equalizing Tube pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Codice prodotto. 10539241 Fornitore Lenz Laborglasinstrumente™ N. del fornitore 04152814
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
For the safe, easy and accurate transfer of fluids in the laboratory. Lenz Laborglasinstrumente™ Cylindrical Dropping Funnels, with Glass Plug are manufactured from durable DURAN™ borosilicate glass.
  • With NS socket and cone
  • With scale (approx. volume/mL)
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifica
Capacità (sistema metrico) 50 mL
Materiale Borosilicate Glass
Cone Size 29/32 NS
Graduato Yes
Include w/o Pressure Equalizing Tube
Product Type Dropping Funnel