Learn More
Lenz Laborglasinstrumente™ Imbuti contagocce cilindrici con tappo in PTFE
Codice prodotto. 10635041
Cambia vista
Click to view available options
Capacità (sistema metrico):
25 ml
50 ml
50 ml
100 ml
100 ml
250 ml
250 ml
500 ml
500 ml
1000 ml
Numero giunto:
14/23 NS
29/32 NS
Include:
Pressure Equalizing Tube
w/o Pressure Equalizing Tube
Dimensione della confezione:
Pezzo
pezzi 2
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) Numero giunto Include Dimensione della confezione
10635041 100 ml 14/23 NS Pressure Equalizing Tube pezzi 2
30564740 1000 ml 29/32 NS Pressure Equalizing Tube Pezzo
10589041 1000 ml 29/32 NS Pressure Equalizing Tube pezzi 2
10047810 1000 ml 29/32 NS w/o Pressure Equalizing Tube pezzi 2
10343001 100 ml 29/32 NS w/o Pressure Equalizing Tube pezzi 2
10500012 250 ml 29/32 NS w/o Pressure Equalizing Tube pezzi 2
10519241 25 ml 14/23 NS w/o Pressure Equalizing Tube pezzi 2
10740651 250 ml 29/32 NS Pressure Equalizing Tube pezzi 2
10149650 500 ml 29/32 NS w/o Pressure Equalizing Tube pezzi 2
10707711 25 ml 14/23 NS Pressure Equalizing Tube pezzi 2
12586229 50 ml 14/23 NS Pressure Equalizing Tube pezzi 2
10590002 50 ml 29/32 NS w/o Pressure Equalizing Tube pezzi 2
10792132 500 ml 29/32 NS Pressure Equalizing Tube pezzi 2
12556229 50 ml 29/32 NS Pressure Equalizing Tube pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Codice prodotto. 10635041 Fornitore Lenz Laborglasinstrumente™ N. del fornitore 04373711
  • ONLINE EXCLUSIVE PRICE
    209.00€
    120.61€ / pezzi 2
    Risparmia 88.39€
    42% Sconto
Spedizione stimata: 23-12-2026
Useful for transferring fluids and handling air-sensitive reagents in a sealed, inert-gas environment. Lenz Laborglasinstrumente™ Pressure Equalizing Dropping Funnels are manufactured from durable DURAN™ borosilicate glass.
  • With scale (approx. volume/mL)
  • With pressure equalizing tube
  • With screwthread retaining device
  • Cylindrical shape with NS socket and cone
  • Corrosion-free and resistant to thermal shock
Specifica
Capacità (sistema metrico) 100 ml
Materiale Vetro borosilicato
Graduato Sì
Include Pressure Equalizing Tube
Numero giunto 14/23 NS
Product Type Imbuto contagocce