missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Cynomolgus CD24 Protein, Fc Tag
457.25€ - 3357.52€
Specifica
Peso molecolare | 29.3 kDa |
---|---|
Sorgente | Cynomolgus CD24, Fc Tag (CD4-C5257) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Lys 54 (Accession # I7GKK1-1). |
Immunogeno | Thr 27 - Lys 54 |
Requisiti di stoccaggio | -20°C |
Alias gene | CD24,CD24A |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17106131
|
ACROBiosystems
CD4-C5257-1MG |
1 mg |
3357.52€
1mg |
Spedizione stimata: 02-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
17196121
|
ACROBiosystems
CD4-C5257-100UG |
100 μg |
457.25€
100µg |
Spedizione stimata: 02-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
CD24 may have a pivotal role in cell differentiation of different cell types. Signaling could be triggered by the binding of a lectin-like ligand to the CD24 carbohydrates, and transduced by the release of second messengers derived from the GPI-anchor. Modulates B-cell activation responses. Promotes AG-dependent proliferation of B-cells, and prevents their terminal differentiation into antibody-forming cells. In association with SIGLEC10 may be involved in the selective suppression of the immune response to danger-associated molecular patterns (DAMPs) such as HMGB1, HSP70 and HSP90. Plays a role in the control of autoimmunity.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 29.3 kDa. The protein migrates as 42-50 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.Specifica
29.3 kDa | |
Thr 27 - Lys 54 | |
CD24,CD24A | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
Cynomolgus CD24, Fc Tag (CD4-C5257) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Lys 54 (Accession # I7GKK1-1). | |
-20°C | |
CD24 | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |