Learn More
ACROBiosystems Cynomolgus / Rhesus macaque CD19 (20-292) Protein, His Tag
535.63€ - 4180.56€
Specifica
Numero di accesso | XP_001103043.1 |
---|---|
Peso molecolare | 32.1 kDa |
Sorgente | Cynomolgus / Rhesus macaque CD19 (20-292), His Tag (CD9-C5221) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 292 (Accession # F7F486-1). |
Immunogeno | Pro 20 - Lys 292 |
Requisiti di stoccaggio | -20°C |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17256120
|
ACROBiosystems
CD9-C5221-1MG |
1 mg |
4180.56€
1mg |
Spedizione stimata: 30-04-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
17246120
|
ACROBiosystems
CD9-C5221-100UG |
100 μg |
535.63€
100µg |
Spedizione stimata: 30-04-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
B-lymphocyte antigen CD19 is also known as CD19 (Cluster of Differentiation 19), is a single-pass type I membrane protein which contains two Ig-like C2-type (immunoglobulin-like) domains. CD19 is expressed on follicular dendritic cells and B cells. In fact, it is present on B cells from earliest recognizable B-lineage cells during development to B-cell blasts but is lost on maturation to plasma cells. It primarily acts as a B cell co-receptor in conjunction with CD21 and CD81. Upon activation, the cytoplasmic tail of CD19 becomes phosphorylated, which leads to binding by Src-family kinases and recruitment of PI-3 kinase. As on T cells, several surface molecules form the antigen receptor and form a complex on B lymphocytes. The (almost) B cell-specific CD19 phosphoglycoprotein is one of these molecules. The others are CD21 and CD81. These surface immunoglobulin (sIg)-associated molecules facilitate signal transduction. On living B cells, anti-immunoglobulin antibody mimicking exogenous antigen causes CD19 to bind to sIg and internalize with it. The reverse process has not been demonstrated, suggesting that formation of this receptor complex is antigen-induced. This molecular association has been confirmed by chemical studies. Mutations in CD19 are associated with severe immunodeficiency syndromes characterized by diminished antibody production. CD19 has been shown to interact with: CD81, CD82, Complement receptor 2, and VAV2.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 32.1 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
XP_001103043.1 | |
Cynomolgus / Rhesus macaque CD19 (20-292), His Tag (CD9-C5221) is expressed from human 293 cells (HEK293). It contains AA Pro 20 - Lys 292 (Accession # F7F486-1). | |
-20°C | |
CD19 | |
Cynomolgus Monkey, Rhesus Macaque | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers & CD Antigens |
32.1 kDa | |
Pro 20 - Lys 292 | |
CD19,B4,CVID3,MGC12802 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |