Learn More
CYP3A7 Antibody, Novus Biologicals™
Codice prodotto. 18535250 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18535250 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18535250 Fornitore Novus Biologicals N. del fornitore NBP162676
Rabbit Polyclonal Antibody
CYP3A7 Polyclonal antibody specifically detects CYP3A7 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifica
Antigene CYP3A7
Applicazioni Western Blot
Classificazione Polyclonal
Concentrazione 0.5 mg/mL
Coniugato Unconjugated
Diluizione Western Blot 1.0 μg/mL
Formulazione PBS, 2% Sucrose
N. accesso geni P24462
Alias gene CYPIIIA7, cytochrome P450, family 3, subfamily A, polypeptide 7, Cytochrome P450-HFLA, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, subfamily IIIA, polypeptide 7, xenobiotic monooxygenase
Specie ospite Rabbit
Immunogeno Synthetic peptides corresponding to CYP3A7(cytochrome P450, family 3, subfamily A, polypeptide 7) The peptide sequence was selected form the middle region of CYP3A7. Peptide sequence KLALVRVLQNFSFKPCKETQIPLKLRFGGLLLTEKPIVLKAESRDETVSG. The peptide sequence for this immunogen was taken from within the described region.
Metodo di purificazione Affinity purified
Quantità 100 μg
Status giuridico RUO
Disciplina di ricerca Lipid and Metabolism
Primario o secondario Primary
ID gene (immissione) 1551
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Forma Purified
Isotype IgG