missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP7B1 Polyclonal Antibody, Invitrogen™
Descrizione
Recommended positive controls: 293T, A431, H1299. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum membrane protein catalyzes the first reaction in the cholesterol catabolic pathway of extrahepatic tissues, which converts cholesterol to bile acids. This enzyme likely plays a minor role in total bile acid synthesis, but may also be involved in the development of atherosclerosis, neurosteroid metabolism and sex hormone synthesis.
Specifica
Specifica
| Antigene | CYP7B1 |
| Applicazioni | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 2 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 40% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Cyp7b1 |
| N. accesso geni | O75881 |
| Alias gene | 24-hydroxycholesterol 7-alpha-hydroxylase; 25/26-hydroxycholesterol 7-alpha-hydroxylase; 25-hydroxycholesterol 7-alpha-hydroxylase; 3-hydroxysteroid 7-alpha hydroxylase; AW261589; CBAS3; CP7B; CYP7B1; Cytochrome P450 7B1; cytochrome P450 family 7 subfamily B member 1; cytochrome P450, 7b1; cytochrome P450, family 7, subfamily b, polypeptide 1; cytochrome P450, subfamily 7B, polypeptide 1; cytochrome P450, subfamily VIIB (oxysterol 7 alpha-hydroxylase), polypeptide 1; D3Ertd552e; hct-1; hippocampal transcript 1 protein; Oxysterol 7-alpha-hydroxylase; SPG5A |
| Simboli geni | Cyp7b1 |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?