Learn More
Cytarabine, TRC
Codice prodotto. 30132486
Cambia vista
Click to view available options
Quantità:
50 mg
100 mg
1 g
Dimensione della confezione:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30132486 100 mg 100mg
30133805 1 g 1g
17981305 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 30132486 Fornitore Toronto Research Chemicals N. del fornitore TRCC998100100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 147-94-4
Formula molecolare C9 H13 N3 O5
Molecular Weight (g/mol) 243.22
Sinonimo Cytarabine, 4-Amino-1-β-d-arabinofuranosylpyrimidin-2(1H)-one, 2(1H)-Pyrimidinone, 4-amino-1-β-D-arabinofuranosyl-, Cytosine, 1-β-D-arabinofuranosyl- (6CI,8CI), 4-Amino-1-β-D-arabinofuranosyl-2(1H)-pyrimidinone, (Arabinofuranosyl)cytosine, 1-(Arabinofuranosyl)cytosine, 1-(β-D-Arabinofuranosyl)cytosine, 1-β-Arabinofuranosylcytosine, 1-β-D-Arabinosylcytosine, 4-Amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one, 4-Amino-1-arabinofuranosyl-2-oxo-1,2-dihydropyrimidine, Ac 1075, Alexan, Ara-C, Ara-cell, Arabinocytosine, Arabinoside C, Arabitin, Aracytidine, Aracytin, Aracytine, Arafcyt, CHX 3311, Citozar, Cyclocide, Cylocide, Cylocide N, Cytarabin, Cytarabinoside, Cytosar, Cytosar U, Cytosine arabinoside, Cytosine β-D-arabinofuranoside, Cytosine β-D-arabinoside, Cytosine-1-β-D-arabinofuranoside, Cytosine-1-β-arabinofuranoside, DepoCyt, DepoCyte, Erpalfa, Iretin, MK 8242, NSC 287459, NSC 63878, SCH 900242, Spongocytidine, Tarabine PFS, U 19920, U 19920A, Udicil, ara-Cytosine
IUPAC Name 4-amino-1-[(2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@@H]2O
Specifica
Materiale o nome chimico Cytarabine
Note sul grado di purezza HPLC
Quantità 100 mg
InChI Formula InChI=1S/C9H13N3O5/c10-5-1-2-12(9(16)11-5)8-7(15)6(14)4(3-13)17-8/h1-2,4,6-8,13-15H,3H2,(H2,10,11,16)/t4-,6-,7+,8-/m1/s1
Peso formulazione 243.0855
Percent Purity >95%