missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cyto-Cal™ Multifluor + Violet Intensity Calibrator

Codice prodotto. 11878293 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
1.5 x 10 particles/mL
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11878293 1.5 x 10 particles/mL Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Contains sodium azide.
Codice prodotto. 11878293 Fornitore Thermo Scientific™ N. del fornitore FC3MV

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Contains sodium azide.

Simultaneously monitor stability and check sensitivity and performance of flow cytometer.

Simultaneously monitor stability and check sensitivity and performance of flow cytometer with the Thermo Scientific™ Cyto-Cal™ Multifluor + Violet Intensity Calibrator.

Features:
  • Stable calibration material for assessing instrument stability and precision in multiple channels
  • Uniform mixture of 3μm particles with four dyes in five different fluorescent intensities
  • Singlet gating not required due to high uniformity of the particles
  • Consists of a single vial of fluorescent particles precisely stained with fluorescent dyes that have optimized intensity and broad emission in up to 13 color channels
  • MESF (Mean Equivalent Soluble Fluorochrome) value provided for each intensity level for estimation of relative fluorescence intensities of labeled cells
  • Can provide relative fluorescence estimations of labeled cells (MEFL), but is not designed for quantitation of fluorochromes on cells.

Includes:

Insert sheet and MSDS

Recommended for:

Flow Cytometry set-up and calibration.

Note:

MEFL is also referred to as Molecules of Equivalent Soluble Fluorochrome (MESF). The MEFL values provided are not traceable to a standard reference material.

TRUSTED_SUSTAINABILITY

Specifica

Additivo 0.05% Tween-20 Dispersant / Surfactant with 2 mM Sodium Azide preservative
Descrizione Beads for assessing instrument stability and precision in multiple channels
Diametro (sistema metrico) 3 μm
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Tipo di colorante Firefli™ Fluorescent Green (488/510nm), Orange (488/575nm), Red (488, 633, 635/700nm) and Blue (405/
Densità particelle 1.06g/cm3
Materiale Polystyrene
N. di test 50
Livello 1 blank and 5 fluorescent
Include Dyed polymer microspheres in water, Package insert sheet
Stabilità Stable for a minimum of 12 months
Quantità 1.5 x 10 particles/mL
Volume (sistema metrico) 2 mL
Uniformità <3%
Product Line Cyto-Cal
Product Type Instrument Calibration
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.