missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cytochrome P450 Pan Polyclonal Antibody
GREENER_CHOICE

Codice prodotto. 15507581
Cambia vista
Click to view available options
Quantità:
100 μL
Dimensione della confezione:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15507581 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15507581 Fornitore Invitrogen™ N. del fornitore PA536232

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 60 kDa. Purity is >95% by SDS-PAGE.

CYP2C9 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. CYP2C9 localizes to the endoplasmic reticulum and its expression is induced by rifampin. The enzyme is known to metabolize many xenobiotics, including phenytoin, tolbutamide, ibuprofen and S-warfarin.
TRUSTED_SUSTAINABILITY

Specifica

Antigene Cytochrome P450 Pan
Applicazioni Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classificazione Polyclonal
Concentrazione 1 mg/mL
Coniugato Unconjugated
Formulazione PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene CYP2C9
N. accesso geni P05181, P05182, P10632, P11712, P33260, P33261, Q05421
Alias gene (R)-limonene 6-monooxygenase; (S)-limonene 6-monooxygenase; (S)-limonene 7-monooxygenase; (S)-mephenytoin hydroxylase associated cytochrome P450; cholesterol 25-hydroxylase; CPC8; CPC9; CPCI; CPCJ; CYP2C; CYP2C10; CYP2C17; CYP2C18; CYP2C19; CYP2C8; CYP2C9; CYPIIC17; CYPIIC18; CYPIIC19; CYPIIC8; CYPIIC9; Cytochrome P450 2C18; cytochrome P450 2C19; Cytochrome P450 2C8; cytochrome P450 2C9; cytochrome P450 family 2 subfamily C member 18; cytochrome P450 family 2 subfamily C member 19; cytochrome P450 family 2 subfamily C member 8; cytochrome P450 family 2 subfamily C member 9; cytochrome P450 form 1; cytochrome P-450 II C; Cytochrome P450 IIC2; cytochrome P450 MP-12; Cytochrome P450 MP-20; Cytochrome P450 MP-4; Cytochrome P450 MP-8; Cytochrome P450 PB-1; cytochrome P-450 S-mephenytoin 4-hydroxylase; cytochrome P450, family 2, subfamily C, polypeptide 18; cytochrome P450, family 2, subfamily C, polypeptide 19; cytochrome P450, family 2, subfamily C, polypeptide 8; cytochrome P450, family 2, subfamily C, polypeptide 9; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 17; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 18; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 19; cytochrome P450, subfamily IIC (mephenytoin 4-hydroxylase), polypeptide 8; cytochrome P450-11A; Cytochrome P450-254C; cytochrome P450-6B/29C; cytochrome P-450MP; Fenbendazole monooxygenase (4'-hydroxylating); flavoprotein-linked monooxygenase; mephenytoin 4'-hydroxylase; mephenytoin 4-hydroxylase; microsomal monooxygenase; MP-12/MP-20; P450 form 1; P450-6B/29C; P450C2C; P450IIC17; P450IIC19; P450IIC9; s-mephenytoin 4-hydroxylase; unspecific monooxygenase; xenobiotic monooxygenase
Simboli geni CYP2C18, CYP2C19, CYP2C8, CYP2C9, CYP2E1
Specie ospite Rabbit
Immunogeno Synthetic peptide corresponding to amino acids 100-150 of human CYP2C8.
Metodo di purificazione Antigen Affinity Chromatography
Quantità 100 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 13106, 1557, 1558, 1559, 1562, 1571, 25086
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Forma Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.