Learn More
Cytiva Cytodex 1 Gamma Microcarriers
Codice prodotto. 15717207 Sfoglia Tutto Cytiva Prodotti
Cambia vista
Click to view available options
Quantità:
30 g
300 g
3 kg
Dimensione della confezione:
300g
30g
3kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15717207 30 g 30g
15727207 300 g 300g
15737207 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 15717207 Fornitore Cytiva N. del fornitore 17548701
Microcarrier for general use in sterilized dry powder form.
  • Cytodex 1 Gamma is designed by substituting a cross-linked dextran matrix with positively charged N, N-diethylaminoethyl (DEAE) groups distributed throughout the matrix.
  • Micro-Carriers Dry Weight: 4.3 × 106
  • Surface Area Dry Weight: 0.44 m2/g
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con (applicazione) Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, diff erentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving celltransfer and studies on uptage of labelled compounds
Quantità 30 g