Learn More
Thermo Scientific™ Cytomat™ 2 C450-LiN Automated Incubator
Product Code. 17880102 Shop All Thermo Scientific Products
Change view
Click to view available options
Temperature Range (Metric):
Ambient +4° to 50°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
17880102 Ambient +4° to 50°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17880102 Supplier Thermo Scientific™ Supplier No. 51033037
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Temperature Range: Ambient +4°C to 50°C; Relative Humidity: <95%
Ensure a safe, reproducible process, unsurpassed temperature uniformity and stability with the next generation of mid-range Thermo Scientific™ Cytomat™ 2 C4xx-LiN series automated incubators for incubation/storage of cell culture applications.
These Cytomat incubators are available in various temperature ranges and gate positions for the hand-off to the lab automation robot. The device can be seamlessly integrated with a mean access time of 12 sec.
With a fully automated thermal decontamination andoptional controlled humidity, the Cytomat 2 C4xx-LiN series provides a new standard in significantly reducing the risk of contamination.
The Cytomat 2 C4xx-LiN series of automated incubators provide an ideal safe environment for storage and incubation of assays. This series of mid-range incubators offer a choice of temperature ranges (4 to 70°C), high humidity up to 95% r.H. stable CO2 conditions, and automated ContraCon decontamination routine without harmful gases.
The external cooling and heating unit is designed to allow the user to create a customized working space for easy system integration into any automated assay system.
Available options
  • Integrated barcode reader (1D/2D) enables sample tracking, barcode transmission, and inventory management.
  • P3-Plate detection in stacker helps to eliminate labware handling errors.
  • HydraSmart Technology for precise humidity control without heat extraction and external water tank to avoid contamination.
  • Right speed setting for the right application with individual speed setting per plate and adjustable incremental speed settings (Slow/Medium/Fast).
  • Flexible positioning inside your lab with 6 possible gate positions including under bench in a single body.
  • Solid Copper inner chamber for natural contamination prevention.
  • Tower Shaker (ToS) for TRUE orbital shaking of applications requiring sample agitation and cells that need to be kept in suspension.
Specifications
Certifications/Compliance CE Marked
Access Time 12 sec.
Chamber Material Copper
Holds 56 Pc. 1536-Well Plates, 42 Pc. 96-Well or 384-Well Plates, 30 Pc. Transwell or Insert Plates, 20 Pc. Deep-Well
Relative Humidity <95%
Temperature Range (Metric) Ambient +4° to 50°C
Type Automated Incubator
Features Incubation and storage unit with temperature range above; Humidity up to 95% r.H. CO2 range of 0-20 Vol. %; Includes decontamination routine-ContraCon
For Use With (Application) Incubation/storage for cell culture applications 4 to 50°C
Model Cytomat 2 C450-LiN 230V 50Hz
Width (English) 22.5 in.
Width (Metric) 572mm
Height (English) 35.6 in.
Height (Metric) 905mm
Depth (English) 20.1 in.
Depth (Metric) 511mm
Weight (Metric) 90kg
Electrical Requirements 230V 50Hz
Voltage 230 V