missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ CytoScan™ 750K Kit Plus
Descrizione
- The complete CytoScan™ 750K Suite includes CytoScan 750K Arrays, CytoScan Reagent Kit, easy-to-use Chromosome Analysis Suite (ChAS) Software, and the proven GeneChip™ Instrument System.
- CytoScan 750K Suite enables cytogenetic researchers to detect and analyze relevant chromosomal aberrations with confidence. CytoScan 750K Suite provides high-resolution coverage of cancer and constitutional genes of interest, along with high-density SNP coverage for loss of heterozygosity (LOH) detection.
- The CytoScan 750K Array provides a cost-effective solution that delivers industry-leading performance. CytoScan 750K Array, reagents, and software were developed with input from the international cytogenetics community to deliver an intuitive platform for fast and reliable detection of chromosomal aberrations.
- CytoScan 750K Suite is designed to save time and money and reduce operator error. The simplified and streamlined assay protocol is aligned with laboratory workflow requirements, making it easier to obtain consistent and high-quality results. Each component passes strict quality control inspection to ensure the highest reproducibility and performance with CytoScan 750K Arrays. The CytoScan protocol can be accomplished in 2.5 days with just seven hours of hands-on time.
- CytoScan 750K Array includes 200,000 gene-centric SNPs to enable confident breakpoint estimation, 5 Mb loss of heterozygosity (LOH) determination and detection of uniparental iso-disomy (UPD) and regions identical-by-descent.
- The popular ChAS Software is tailored to cytogenetic research analysis and reporting. The software is available free of charge as part of the complete CytoScan 750K Suite.
- To install ChAS onto one or more analysis workstations, download the ChAS Software zip file, extract the zip archive, and install by following the instructions in the Installation Instructions file.
- GeneChip array-specific GeneChip Command Console™ Software plug-in package is required for each array.
- Customer training is required for CytoScan 750K assay for both new and existing customers. Different products are available based upon your experience level; please contact your regional sales distributor for more information. Also, see our CytoScan 750K Training Kit (US and Canada Only).
Specifica
Specifica
| Tipo | 750K Kit Plus 96 |
| Matrice | Cytogenetics, Copy Number |
| Number of Arrays | 96 arrays |
| Formato | Genechip Probe Array |
| Specie | Human |
| Number of Markers | 750,000 markers |
| Quantità | 96 arrays |
For Research Use Only. Not for use in diagnostic procedures.
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?