missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Glutamine, 99+%

Catalog Number p-7041939
28.46 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
1 g
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5959-95-5
C5H10N2O3
146.146
MFCD00065607
ZDXPYRJPNDTMRX-GSVOUGTGSA-N
d-glutamine, h-d-gln-oh, d-2-aminoglutaramic acid, l +-glutamine, 2r-2-amino-4-carbamoylbutanoic acid, r-2,5-diamino-5-oxopentanoic acid, d-glutamic acid 5-amide, d-glutamin, unii-63hb36ca2y, d-gln
145815
CHEBI:17061
(2R)-2,5-diamino-5-oxopentanoic acid
C(CC(=O)N)C(C(=O)O)N
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5959-95-5
C5H10N2O3
146.146
MFCD00065607
ZDXPYRJPNDTMRX-GSVOUGTGSA-N
d-glutamine, h-d-gln-oh, d-2-aminoglutaramic acid, l +-glutamine, 2r-2-amino-4-carbamoylbutanoic acid, r-2,5-diamino-5-oxopentanoic acid, d-glutamic acid 5-amide, d-glutamin, unii-63hb36ca2y, d-gln
145815
CHEBI:17061
(2R)-2,5-diamino-5-oxopentanoic acid
C(CC(=O)N)C(C(=O)O)N

Kinase inhibitor; has antivasospastic properties

Kinase inhibitor; has antivasospastic properties

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico D-Glutamine
Colore White
Beilstein 1723796
Peso formulazione 146.14
Percent Purity ≥99%
Quantità 1 g
Forma fisica Powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'