Learn More
MedChemExpress D-Luciferin sodium
Specifica
Specifica
| CAS | 103404-75-7 |
| Solubilità | H2O : 250 mg/mL (826.99 mM∣Need ultrasonic) ∣DMSO : 100 mg/mL (330.80 mM∣Need ultrasonic) |
| Sinonimo | D-(-)-Luciferin sodium∣Firefly luciferin sodium∣Beetle Luciferin sodium |
| Content And Storage | 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light) |
| Quantità | 10 mg |
| Peso molecolare | 302.3 g/mol |
| Product Type | Dye Reagents |
| Purity or Quality Grade | 98.0% |
| Shipping Condition | Room temperature |