missing translation for 'onlineSavingsMsg'
Learn More

Abcam Dansyl Chloride, Fluorogenic Amine Reactive Reagent, Solid, 1 g

Codice prodotto. 32108842 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
1 g
Dimensione della confezione:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32108842 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 32108842 Fornitore Abcam N. del fornitore ab1463971g
 Articolo soggetto a restrizioni
Spedizione stimata: 28-10-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Fluorogenic primary amine reactive reagent for N-terminal derivatization of amino acids and peptides.

  • Molecular Weight: 269.75 g/mol
  • Purity: >95%
  • Form: Solid
  • Applications: Fluorogenic primary amine reactive reagent for N-terminal derivatization of amino acids and peptides, and for polyamine detection by reverse phase HPLC and other analytical procedures
  • Fluorescence: λex 337 nm; λem 492 nm in chloroform (after derivatization with hexylamine)
  • Composition: 100% Dansyl chloride
  • CAS Number: 605-65-2

The product is subject to the following: Abcam Restricted Use Statement

Specifica

Product Type Protein Labeling Reagent
Tampone 100% Dansyl chloride
Content And Storage Store at -20°C.
Da utilizzare con (apparecchiatura) HPLC
Forma Solid
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Substrato Primary amine
Target Molecule Primary Amine
Metodo di rilevamento Fluorescence
Formulazione 100% Dansyl chloride
Quantità 1 g
Da utilizzare con (applicazione) N-terminal derivatization of amino acids and peptides, polyamine detection by reverse phase HPLC, analytical procedures
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.