Learn More
Motic™ Darkfield Attachment with Iris Diaphragm, SMZ-171
Product Code. 15475522
Change view
Click to view available options
Includes:
Iris diaphragm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
15475522 Iris diaphragm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15475522 Supplier Motic™ Supplier No. 1101000901781
  • ONLINE EXCLUSIVE PRICE
    103.00€
    91.63€ / pezzi 1
    Risparmia 11.37€
    11% Sconto
 Restricted Item
Spedizione stimata: 25-01-2027
Easy-to-use darkfield insert for transmitted light. Motic™ Darkfield Attachment with Iris Diaphragm, SMZ-171 is mounted on top of any transmitted stand light exit.
  • For any transmitted light stand (halogen or LED).
  • Iris diaphragm allows you to vary the illumination aperture according to the objective numerical aperture.
Specifications
Type Darkfield Attachment
For Use With (Equipment) Halogen or LED transmitted light stand, SMZ-171 Microscopes
Includes Iris diaphragm