missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phosphorylating Phosphoramidite, standard grade, 28-400 finish bottle

Product Code. 17987102 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
0.5 g
Unit Size:
0.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17987102 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17987102 Supplier Thermo Scientific™ Supplier No. 27179410
 Restricted Item
In stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phosphorylating phosphoramidite can be utilized to achieve phosphorylation of the 5' oligo terminus (also known as Phosphate-ON or Chemical Phosphorylation Reagent (CPR)).

Phosphorylating phosphoramidite can be utilized to achieve phosphorylation of the 5' oligo terminus (also known as Phosphate-ON or Chemical Phosphorylation Reagent (CPR)). Chemical phosphorylation is a cost-effective substitute to enzymatic methods (via T4 kinase and ATP), allowing efficient introduction of terminal phosphate groups. Oligos with a 5'-phosphate are conducive to a number of various applications utilizing the ligation of oligos to each other, such as NGS adapters, cloning, and gene construction. This product is supplied in a 30 mL amber bottle with a 28-400 Finish neck style, which is compatible with most MerMade Systems of oligonucleotide synthesizers.

Features:
• Full chemical name: 2-[2-(4,4-Dimethoxytrityloxy)ethylsulfonyl]ethyl-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula: C34H45F3N2O7PS
• Formula weight: 656.66 g/mol
• HPLC Purity: ≥95.0%

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) MerMade Synthesizers
Product Type Phosphorylating CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type None (Modifier)
Shipping Condition Room Temperature
Purity 95%
Quantity 0.5 g
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type None (Modifier)
Form Powder
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.