missing translation for 'onlineSavingsMsg'
Learn More

Daxx Antibody (DAXX-01), Biotin, Novus Biologicals™

Codice prodotto. 18565922 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18565922 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18565922 Fornitore Novus Biologicals N. del fornitore NB500491B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Daxx Monoclonal antibody specifically detects Daxx in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigene Daxx
Applicazioni Western Blot, Immunocytochemistry/Immunofluorescence
Classificazione Monoclonal
Clone DAXX-01
Coniugato Biotin
Formulazione PBS
Alias gene BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246
Specie ospite Mouse
Immunogeno Recombinant C-terminal part (aa 558-740) of human Daxx.
Metodo di purificazione Protein A purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Apoptosis, Cancer, Tumor Suppressors
Primario o secondario Primary
ID gene (immissione) 1616
Target Species Human
Content And Storage Store at 4°C in the dark.
Forma Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.