Learn More
DBZ, Tocris Bioscience™
γ-secretase inhibitor; inhibits Notch pathway
Marca: Tocris Bioscience 4489/50
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Inhibitor of γ-secretase. Blocks Notch cleavage (IC50< 2 nM) in a cell-based assay; inhibits the Notch pathway. Stimulates the formation of human iPSCs from human keratinocytes.Specifica
| DBZ | |
| 209984-56-5 | |
| >98% | |
| 50 mg |