Learn More
DCANP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561706
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: KSLKSSDPWH PPSLSPNSWN RQAGFRAWSS HLISLSLTCS DSQSRRVSSS QQPPLHSLSS HRRAAHVPE Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 32%.
DCANP1 Polyclonal antibody specifically detects DCANP1 in Human samples. It is validated for Immunohistochemistry.Specifica
DCANP1 | |
Polyclonal | |
Unconjugated | |
DCANP1 | |
C5orf20; DCANP1; DCNP1; dendritic cell associated nuclear protein; dendritic cell nuclear protein 1; Dendritic cell nuclear protein-1; dendritic cell-associated nuclear protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140947 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TF63 | |
DCANP1 | |
Recombinant protein corresponding to Human DCANP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only