missing translation for 'onlineSavingsMsg'
Learn More

delta-1,2,3,4,5,6-Hexachlorocyclohexane, TRC

Codice prodotto. 30401748
Cambia vista
Click to view available options
Quantità:
5 mg
Dimensione della confezione:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30401748 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Product Code. 30401748 Supplier Toronto Research Chemicals Supplier No. TRCH2908005MG

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico delta-HCH
CAS 319-86-8
Formula molecolare C6 H6 Cl6
Quantità 5 mg
Sinonimo (1α,2α,3α,4β,5α,6β)-1,2,3,4,5,6-Hexachlorocyclohexane, Delta-BHC, Δ-1,2,3,4,5,6-Hexachlorocyclohexane, Δ-666, Δ-BHC, Δ-Benzene hexachloride, Δ-HCH, Δ-Hexachlorocyclohexane, Δ-Lindane
InChI Formula InChI=1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4+,5-,6-
SMILES Cl[C@@H]1[C@H](Cl)[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@H]1Cl
IUPAC Name 1,2,3,4,5,6-hexachlorocyclohexane
Molecular Weight (g/mol) 290.83
Peso formulazione 287.8601
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.