Learn More
Delta2-Ceftazidime (>80%), TRC
Specifications
Specifications
| CAS | 1000980-60-8 |
| Molecular Weight (g/mol) | 546.58 |
| Quantity | 25 mg |
| Shipping Condition | Room Temperature |
| Synonym | Pyridinium, 1-[[(6R,7R)-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-3-en-3-yl]methyl]-, inner salt∣Ceftazidime Delta-3-Isomer |