Learn More
Alla France™ Density Hydrometer
Product Code. 11703413
Change view
Click to view available options
Specific Gravity Range:
0.7 to 0.750 g/mL
0.750 to 0.800 g/mL
0.800 to 0.850 g/mL
0.850 to 0.900 g/mL
0.900 to 0.950 g/mL
0.950 to 1.000 g/mL
1.000 to 1.050 g/mL
1.050 to 1.100 g/mL
1.100 to 1.150 g/mL
1.150 to 1.200 g/mL
1.200 to 1.250 g/mL
1.250 to 1.300 g/mL
1.300 to 1.350 g/mL
1.350 to 1.400 g/mL
1.400 to 1.450 g/mL
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Specific Gravity Range Unit Size
11703413 1.050 to 1.100 g/mL Each
11703403 0.7 to 0.750 g/mL Each
11743403 0.750 to 0.800 g/mL Each
11753403 0.800 to 0.850 g/mL Each
11763403 0.850 to 0.900 g/mL Each
11773403 0.900 to 0.950 g/mL Each
11783403 0.950 to 1.000 g/mL Each
11793403 1.000 to 1.050 g/mL Each
11713413 1.100 to 1.150 g/mL Each
11723413 1.150 to 1.200 g/mL Each
11733413 1.200 to 1.250 g/mL Each
11743413 1.250 to 1.300 g/mL Each
11773413 1.400 to 1.450 g/mL Each
11753413 1.300 to 1.350 g/mL Each
11763413 1.350 to 1.400 g/mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 11703413 Supplier Alla France™ Supplier No. 3500LC105/20/Mqp
  • ONLINE EXCLUSIVE PRICE
    86.38€
    33.97€ / pezzi 1
    Risparmia 52.41€
    60% Sconto
 Restricted Item
Spedizione stimata: 07-12-2026
Use these thermometers for a variety of applications. Alla France™ Density Hydrometers are fully marked with all the reference norms. Benchmark engraved on the stem to show any displacement of the scale.
  • Density Hydrometers (g/mL)
  • ISO, BS, DIN, NF... MRA Traceable
  • General Purpose at 20°C
  • Reading below meniscus
  • Systematically delivered with a Traceability and Conformity Certificate
  • On request, can be supplied with "EU" Traceable Calibration Certificate or "EO" Official Calibration Certificate
  • Calibrated according to the following surface tensions:
  • from 0.600 to 1.000 g/mL= 16 at 35 mN/m (low tension)
  • from 1.000 to 1.300 g/mL= 55 mN/m (medium tension)
  • from 1.300 to 2.000 g/mL= 75 mN/m (high tension)
Specifications
Quantity 1Each
Type Density Hydrometer
Accuracy ±0.0005g/mL
Length (Metric) 335 mm
Specific Gravity Range 1.050 to 1.100 g/mL