Learn More
Alla France™ Idrometro di densità
Codice prodotto. 11783403
Cambia vista
Click to view available options
Intervallo di gravità specifica:
0.7 to 0.750 g/mL
0.750 to 0.800 g/mL
0.800 to 0.850 g/mL
0.850 to 0.900 g/mL
0.900 to 0.950 g/mL
0.950 to 1.000 g/mL
1.000 - 1.050 g/ml
1.050 - 1.100 g/ml
1.100 to 1.150 g/mL
1.150 to 1.200 g/mL
1.200 to 1.250 g/mL
1.250 to 1.300 g/mL
1.300 to 1.350 g/mL
1.350 to 1.400 g/mL
1.400 to 1.450 g/mL
Dimensione della confezione:
Pezzo
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Intervallo di gravità specifica Dimensione della confezione
11783403 0.950 to 1.000 g/mL Pezzo
11703403 0.7 to 0.750 g/mL Pezzo
11743403 0.750 to 0.800 g/mL Pezzo
11753403 0.800 to 0.850 g/mL Pezzo
11763403 0.850 to 0.900 g/mL Pezzo
11773403 0.900 to 0.950 g/mL Pezzo
11793403 1.000 - 1.050 g/ml Pezzo
11703413 1.050 - 1.100 g/ml Pezzo
11713413 1.100 to 1.150 g/mL Pezzo
11723413 1.150 to 1.200 g/mL Pezzo
11733413 1.200 to 1.250 g/mL Pezzo
11743413 1.250 to 1.300 g/mL Pezzo
11773413 1.400 to 1.450 g/mL Pezzo
11753413 1.300 to 1.350 g/mL Pezzo
11763413 1.350 to 1.400 g/mL Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Codice prodotto. 11783403 Fornitore Alla France™ N. del fornitore 3500LC095/20/Bqp
  • ONLINE EXCLUSIVE PRICE
    85.49€
    32.48€ / pezzi 1
    Risparmia 53.01€
    62% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 18-11-2026
Use these thermometers for a variety of applications. Alla France™ Density Hydrometers are fully marked with all the reference norms. Benchmark engraved on the stem to show any displacement of the scale.
  • Density Hydrometers (g/mL)
  • ISO, BS, DIN, NF... MRA Traceable
  • General Purpose at 20°C
  • Reading below meniscus
  • Systematically delivered with a Traceability and Conformity Certificate
  • On request, can be supplied with "EU" Traceable Calibration Certificate or "EO" Official Calibration Certificate
  • Calibrated according to the following surface tensions:
  • from 0.600 to 1.000 g/mL= 16 at 35 mN/m (low tension)
  • from 1.000 to 1.300 g/mL= 55 mN/m (medium tension)
  • from 1.300 to 2.000 g/mL= 75 mN/m (high tension)
Specifica
Quantità 1Each
Tipo Density Hydrometer
Accuracy ±0.0005g/mL
Lunghezza (sistema metrico) 335 mm
Intervallo di gravità specifica 0.950 to 1.000 g/mL