Descrizione
This ensures that the sampler penetrates to the same desired depth each time it is inserted. The penetration depth is thus fixed and can be precisely recorded in a test report or sampling instructions, for example. The depth stop can also prevent unintentional piercing during sampling.
- Enables standardized and therefore reproducible sampling
- Very good clamping effect due to C-clamping screw
- Available for 25/50 mm diameter, suitable for many sampler lances
Specifica
Specifica
| Product Type | Sampler Insert |
| Materiale | Stainless Steel (V4A) |
| Autoclavabile | Autoclavable |