Learn More
Des-glutamic Acid-(S)-Folinic Acid, TRC
Codice prodotto. 30324901
Cambia vista
Click to view available options
Quantità:
2.5 mg
Dimensione della confezione:
2.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30324901 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30324901 Fornitore Toronto Research Chemicals N. del fornitore TRCA1558402.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico (6S)-5-Formyltetrahydropteroic Acid
CAS 944737-05-7
Formula molecolare C15 H16 N6 O4
Quantità 2.5 mg
Sinonimo 4-[[[(6S)-2-Amino-5-formyl-3,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl]methyl]amino]benzoic Acid, Benzoic acid, 4-[[[(6S)-2-amino-5-formyl-3,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl]methyl]amino]-, 4-[[[(6S)-2-Amino-5-formyl-3,4,5,6,7,8-hexahydro-4-oxo-6-pteridinyl]methyl]amino]benzoic acid, (6S)-5-Formyl-5,6,7,8-tetrahydropteroic acid, (6S)-5-Formyltet
InChI Formula InChI=1S/C15H16N6O4/c16-15-19-12-11(13(23)20-15)21(7-22)10(6-18-12)5-17-9-3-1-8(2-4-9)14(24)25/h1-4,7,10,17H,5-6H2,(H,24,25)(H4,16,18,19,20,23)/t10-/m0/s1
SMILES NC1=NC(=O)C2=C(NC[C@H](CNc3ccc(cc3)C(=O)O)N2C=O)N1
IUPAC Name 4-[[(6S)-2-amino-5-formyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl]methylamino]benzoic acid
Molecular Weight (g/mol) 344.325
Peso formulazione 344.123