Learn More
Desamino Imiquimod N-Oxide, TRC
Codice prodotto. 30363692
Cambia vista
Click to view available options
Quantità:
250 mg
Dimensione della confezione:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30363692 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30363692 Fornitore Toronto Research Chemicals N. del fornitore TRCD288305250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Desaminoimiquimod N-Oxide
CAS 99010-63-6
Formula molecolare C14 H15 N3 O
Quantità 250 mg
Sinonimo 1H-Imidazo[4,5-c]quinoline, 1-(2-methylpropyl)-, 5-oxide, 1-Isobutyl-1H-imidazo[4,5-c]quinoline N-oxide, 1-Isobutyl-1H-imidazo[4,5-c]quinoline 5-Oxide, Desaminoimiquimod N-Oxide
InChI Formula InChI=1S/C14H15N3O/c1-10(2)7-16-9-15-12-8-17(18)13-6-4-3-5-11(13)14(12)16/h3-6,8-10H,7H2,1-2H3
SMILES CC(C)Cn1cnc2c[n+]([O-])c3ccccc3c12
IUPAC Name 1-(2-methylpropyl)-5-oxidoimidazo[4,5-c]quinolin-5-ium
Molecular Weight (g/mol) 241.29
Peso formulazione 241.1215