Learn More
R&D Systems™ Detergent Reagent
Codice prodotto. 30001047
Cambia vista
Click to view available options
Quantità:
250 mL
Dimensione della confezione:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30001047 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30001047 Fornitore R&D Systems™ N. del fornitore 48902502
Please to purchase this item. Need a web account? Register with us today!
R&D Systems™ Detergent Reagent is supplied ready to use and is for use with cell proliferation assays.
  • Do not use past expiration date.
  • Supplied ready for use.
  • If the Detergent Reagent has been stored at 4°C, warm the bottle for 5 minutes at 37°C then invert gently while mixing to avoid frothing.
  • Contains SDS which is an irritant. In case of contact, immediately flush eyes or skin with copious amounts of water. If swallowed, wash out mouth with water provided person is conscious.
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage Store the unopened product at room temperature
Quantità 250 mL