Learn More
Devimistat, MedChemExpress
Codice prodotto. 16469117
Encompass_Preferred
Cambia vista
Click to view available options
Quantità:
5 mg
10 mg
50 mg
100 mg
Dimensione della confezione:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16469117 5 mg 5mg
16452627 50 mg 50mg
16436776 100 mg 100mg
16426538 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 16469117 Fornitore Medchem Express N. del fornitore HY15453
MedChemExpress Devimistat (CPI-613) is a mitochondrial metabolism inhibitor. Devimistat is a lipoic acid antagonist that abrogates mitochondrial energy metabolism to induce apoptosis in various cancer cells.
Devimistat (CPI-613) is a mitochondrial metabolism inhibitor. Devimistat is a lipoic acid antagonist that abrogates mitochondrial energy metabolism to induce apoptosis in various cancer cells.
Specifica
Materiale o nome chimico Devimistat
Forma fisica Solid
CAS 95809-78-2
Colore White
Quantità 5 mg
Note sul grado di purezza Research
Formula molecolare C22H28O2S2
Da utilizzare con (applicazione) Cancer-programmed cell death
Sinonimo CPI-613
Informazioni di solubilità DMSO : 100 mg/mL (257.34 mM; Need ultrasonic)
SMILES O=C(O)CCCCC(SCC1=CC=CC=C1)CCSCC2=CC=CC=C2
Molecular Weight (g/mol) 388.59
Peso formulazione 388.59
Percent Purity 98.0%
Grado Research
Research purposes only